Fisher Scientific™ Traceable™ Excursion-Trac™ Datalogging Thermometers: PROMO OFFER
PROMOTIONAL OFFER: Buy a Fisher Scientific Traceable Excursion-Trac Datalogging Thermometer and get a Fisher Scientific Traceable Radio Atomic Clock (15-076-6) at no additional cost
 View more versions of this product
Catalog No. 15-081-123PMO
  Check Availability
Add to Cart
    Specifications
    Description 1 Bottle Probe
    Product Type Datalogging Thermometer,Datalogging Thermometer
    Includes Stand, Velcro magnetic strips, wall mount, batteries
    Temperature Range (Metric) -50 to -70°C
    Accuracy ±0.25°C
    Measurement Time User-defined timing interval: Yes
    Probe Type 1 Bottle Probe
    Certifications/Compliance ISO 17025, A2LA, NIST
    Resolution 0.01°
    Data Storage Most recent 525,600 temperature observations
  • Description & Specifications
Related Products
Product Content Feedback