Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ One-Shot™ (CRM) pH Buffer Standards PROMO

Fisherbrand™ Traceable™ One-Shot™ (CRM) pH Buffer Standards perform accurate, reproducible pH calibrations with color-coded bottles.

$18.70

Specifications

Additional Information Accuracy of pH: ±0.010 at 25°C
Chemical Name or Material Buffer Standards One-Shot (CRM)
NIST Traceable Certified and Traceable™
View More Specs

Products 3
Catalog Number pH Packaging Price Quantity & Availability  
Catalog Number pH Packaging Price Quantity & Availability  
15-078-287FBS
View Documents
10.012 100 mL One-Shot (6/pk) Pack of 6 for $18.70
Add to cart
 
15-078-286FBS
View Documents
7.00 100 mL One-Shot (6/pk) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
15-078-285FBS
View Documents
4.005 100 mL One-Shot (6/pk) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
  • 100% compatible with all makes of pH instruments and electrodes
  • Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
  • Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
  • Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
  • Supplied withindividual temperature chart, traceability information and Traceable™ Certificate
Specifications

Specifications

Accuracy of pH: ±0.010 at 25°C
Buffer Standards One-Shot (CRM)
Certified and Traceable™
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit