Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Rapid-Response Folding Stainless Steel Thermometer

Fisherbrand Traceable™ Rapid-Response Folding Stainless Steel Thermometer is a wide range thermometer that covers 99 percent of routine temperature checks with a rapid reponse of 2 seconds within 1 degree.

 View more versions of this product

Catalog No. 15-078-301

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Features:

  • Rates at IP65 splash resistance
  • Protects against low-pressure jets of water, such as splashes from a tap
  • Built-in magnetic backing to allow for convenient storage
  • Automatically enters sleep mode after 60 seconds to conserve battery life
  • Water/splash resistant
  • Safe Anti-microbial surface
  • Magnetic Backing
  • Sleep Mode capabilities
  • Fold-up, protected, stainless-steel probe permits ultimate portability can position at any angle
  • Big-digit, 2-inch-high display allows easy to read temperatures.

Compliance

ISO 17025, A2LA, NIST

Warranty and Services

1 year

Specifications

Specifications

-40°C to +250°C
Approx. 4000 hr.
0.1°
ISO 17025, NIST
1.75 in.
Stainless Steel
Stainless Steel
0.75 in.
±0.5°C between 20°C and 250°C, otherwise ±1°C
AAA
0>°C, 100°C and 200°C
6in x 0.75in x 1.75in
Traceable Certificate, Battery
Deluxe
2.375 in.
6 in.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit