Learn More
Fisherbrand™ TraceableOne™ 40 Pack Single-User Datalogger
Use logger right out of the box to track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals and food items during transit. Fisherbrand™ TraceableOne™ 40 Pack Single-User Datalogger allows for monitoring materials every step of the way.
$1040.00
Specifications
Accuracy (Temperature) ±0.5°C from -20° to 40°C, otherwise ±1°C
Calibration Yes
Product Type Single-Use Datalogger
Temperature Range (Metric) −30°C to 70°C
For Use With (Application) Refrigerated transportation samples
View More Specs
Products 8
Catalog Number Alarm Types Data Memory Price Quantity & Availability  
Catalog Number Alarm Types Data Memory Price Quantity & Availability  
15-079-684
View Documents
Preprogrammed -20°C to -15°C Logging Interval of 3 minute, logging duration of 30 days Pack of 40 for $1,040.00
Add to cart
15-079-681
View Documents
Preprogrammed 2°C to 8°C Logging Interval of 1 minute, logging duration of 10 days Pack of 40 for $1,040.00
Add to cart
15-079-682
View Documents
Preprogrammed -20°C to -15°C Logging Interval of 1 minute, logging duration of 10 days Pack of 40 for $1,040.00
Add to cart
15-079-685
View Documents
Preprogrammed 2°C to 8°C Logging Interval of 6 minute, logging duration of 60 days Pack of 40 for $1,040.00
Add to cart
15-079-686
View Documents
Preprogrammed -20°C to -15°C Logging Interval of 6 minute, logging duration of 60 days Pack of 40 for $1,040.00
Add to cart
15-079-683
View Documents
Preprogrammed 2°C to 8°C Logging Interval of 3 minute, logging duration of 30 days Pack of 40 for $1,040.00
Add to cart
15-079-688
View Documents
Preprogrammed -20°C to -15°C Logging Interval of 10 minute, logging duration of 90 days Pack of 40 for $1,040.00
Add to cart
15-079-687
View Documents
Preprogrammed 2°C to 8°C Logging Interval of 10 minute, logging duration of 90 days Pack of 40 for $1,040.00
Add to cart
Description
Description
  • Logger generates PDF report
  • Download directly to PC
  • No Software required
  • Compact
  • Single-use
  • Flashing Red and Green LED's indicate if logger is recording data, has stopped recording and if an alarm has been triggered
  • Press start to begin recording crucial temperature data, 30 minute time delay before logging begins
Specifications
Specifications
±0.5°C from -20° to 40°C, otherwise ±1°C
Single-Use Datalogger
Refrigerated transportation samples
IP67
Battery-Powered
0.1
Yes
−30°C to 70°C
Batch-calibrated Traceable certificate
8.89 cm
No software required, download data as PDF report
40/Pk