Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Dibenz[a,h]anthracene certified reference material, MilliporeSigma™ Supelco™

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Manufacturer:  MilliporeSigma Supelco 918611G

 View more versions of this product

Catalog No. 11-101-7935


Add to cart

Description

Description

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications

Specifications

53-70-3
MFCD00003708
278.35
TraceCERT
262°C to 265°C
1 g
Certified Reference Material
C22H14
1,2:5,6-Dibenzanthracene
Neat
Bottomless Glass Bottle with Inserted Fused Cone
524°C
TraceCERT
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : HN2625000

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC