Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Dibenz[a,h]anthracene certified reference material, MilliporeSigma™ Supelco™

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

$97.50 - $1731.00

Specifications

Product Line TraceCERT
Grade TraceCERT
Linear Formula C22H14
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9202
View Documents
MilliporeSigma Supelco
9186110MG
10 mg
Each for $97.50
Add to cart
 
11-101-7935
View Documents
MilliporeSigma Supelco
918611G
1 g
Each for $1,731.00
Add to cart
 
Description

Description

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications

Specifications

TraceCERT
TraceCERT
C22H14
Neat
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : HN2625000

SDS
Product Certifications

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit