Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Indeno[1,2,3-cd]pyrene, MilliporeSigma™ Supelco™

$113.30

Specifications

Product Line TraceCERT
Grade TraceCERT
Linear Formula C22H12
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3421
View Documents
MilliporeSigma Supelco
9437710MG
10 mg
Each for $113.30
Add to cart
 
11-101-3420
View Documents
MilliporeSigma Supelco
943771G
1 g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Environmental
Specifications

Specifications

TraceCERT
TraceCERT
C22H12
Neat
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit