Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TraceCERT™ Maleate Standard for IC, 1,000 mg/L Maleate in Water, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
100 mL
Description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the label.

Specifications
Specifications
CAS | 110-16-7 |
Product Line | TraceCERT |
Purity Grade Notes | 1000 mg/L maleate aq. soln., prepared with maleic acid and water; filtered through a 0.2 mum membrane filter and stabilized with sodium azide |
Concentration | 1000 mg/L maleate aq. soln. |
Type | Certified Reference Material |
Quantity | 100 mL |
Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction