Learn More
TraceCERT™ TCL Phenols Mix, 2000 μg/mL, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.
Supplier: MilliporeSigma Supelco 48904
Specifications
UN 1593 6.1 / PGIII | |
Ampule | |
Not Applicable |
Trace CERT™ | |
1 mL | |
Organic Pollutant Standards |
Safety and Handling
H315 - H319 - H336 - H351 - H412
Recommended Storage : 2°C to 8°C
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC