Learn More
Thermo Scientific™ TraceGOLD™ TG-200MS GC Columns
Resolve compounds that phenyl and cyano phases cannot using a low-bleed GC column with an exceptionally inert phase.
$790.50 - $1275.00
Specifications
Length (Metric) 30 m
For Use With (Application) ECD, NPD, and GC/MS for Alcohols and Ketones, ECD, NPD, and GC/MS for Fluorocarbons, ECD, NPD, and GC/MS for Glycols, ECD, NPD, and GC/MS for Silanes, ECD, NPD, and GC/MS for Solvents
Stationary Phase TraceGOLD™ TG-200 MS
USP Type G6
View More Specs
Products 8
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Film Thickness Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Film Thickness Price Quantity & Availability  
26-084-1420
Accessories View Documents
Thermo Scientific™
260841420
0.25 mm 30 m 0.25 µm Each for $790.50
Add to cart
26-084-1550
View Documents
Thermo Scientific™
260841550
0.32 mm 60 m 0.25 μm Each for $1,275.00
Add to cart
26-084-1430
View Documents
Thermo Scientific™
260841430
0.32 mm 30 m 0.25 µm Each for $889.50
Add to cart
03-052-624
View Documents
Thermo Scientific™
260842980
0.53 mm 30 m 1 μm Each for $811.00
Add to cart
26-084-2240
View Documents
Thermo Scientific™
260842240
0.32 mm 30 m 0.5 µm Each for $889.50
Add to cart
26-084-2960
View Documents
Thermo Scientific™
260842960
0.25 mm 30 m 1 µm Each for $790.50
Add to cart
26-084-2970
View Documents
Thermo Scientific™
260842970
0.32 mm 30 m 1 µm Each for $889.50
Add to cart
26-084-2230
View Documents
Thermo Scientific™
260842230
0.25 mm 30 m 0.5 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Resolve compounds that phenyl and cyano phases cannot using the Thermo Scientific™ TraceGOLD™ TG-200MS GC Column. The TraceGOLD TG-200MS column has an exceptionally inert phase with selectivity and elution order optimized for difficult separations.
Engineered for Superior Performance
  • Polar phase, trifluoropropylmethyl polysiloxane solid phase to resolve compounds that phenyl and cyano phases cannot
  • Outstanding thermal stability and low bleed
  • Suitable for use with sensitive detectors including electrochemical detectors, nitrogen phosphorus detectors and mass spectrometers
  • Appropriate for use as a confirmation GC column
Applications:
  • Solvents
  • Fluorocarbons
  • Alcohols and ketones
  • Silanes
  • Glycols
A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • This low-bleed, stable column produces consistent results, and the low baseline noise enables improved limits of detection with enhanced resolution of low-level analytes.
  • Minimizing column bleed extends the column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Recommended For:
  • Solvents
  • Fluorocarbons
  • Alcohols and ketones
  • Silanes
  • Glycols
  • Alternative Uses:
  • Herbicides
  • Organochlorine pesticides
  • Phenols
  • Similar To:
  • Rtx™-200MS
  • DB-200
  • DB-210
Specifications
Specifications
30 m
ECD, NPD, and GC/MS for Alcohols and Ketones, ECD, NPD, and GC/MS for Fluorocarbons, ECD, NPD, and GC/MS for Glycols, ECD, NPD, and GC/MS for Silanes, ECD, NPD, and GC/MS for Solvents
TraceGOLD™ TG-200 MS
G6
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.