Learn More
Thermo Scientific™ TraceGOLD™ TG-OCP I and II GC Columns Available on GSA/VA Contract for Federal Government customers only.
Analyze organochlorine pesticides and herbicides with ultra low bleed, superior sensitivity, excellent peak shape and the highest levels of reproducibility.
Manufacturer:  Thermo Scientific™ 260775690
 View more versions of this product
Catalog No. 26-077-5690
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Analyze organochlorine pesticides and herbicides with ultralow bleed, superior sensitivity, excellent peak shape and the highest levels of reproducibility using Thermo Scientific? TraceGOLD? TG-OCP I and TG-OCP II gas chromatography columns.
Description & Specifications
Specifications
GC/MS
0.18mm
20m
20m x 0.18mm x 0.14um
340°C
0.14μm
TraceGOLD™ TG-OCP II