Learn More
Thermo Scientific™ TraceGOLD™ TG-OPP I and II GC Columns
Analyze organophosphorus pesticides with ultra-low bleed, superior inertness and consistent reproducibility.
Manufacturer:  Thermo Scientific™ 260752250
 View more versions of this product
Catalog No. 26-075-2250
Add to cart
Description
Description
Analyze organophosphorus pesticides with ultralow bleed, superior inertness, and consistent reproducibility using Thermo Scientific™ TraceGOLD™ TG-OPP I and II GC Columns.This column's low bleed delivers excellent signal-to-noise ratio, sensitivity, and mass spectral integrity. Expect full separation of organophosphorus pesticides with a fast analysis time. TraceGOLD TG-OPP I and II GC Columns are ideal for U.S. EPA methods 8141A.
Specifications
Specifications
GC/MS
0.53 mm
TraceGOLD TG-OPP II
330°C
30 m