Learn More
Thermo Scientific™ TraceGOLD™ TG-OPP I and II GC Columns Available on GSA/VA Contract for Federal Government customers only.
Analyze organophosphorus pesticides with ultra-low bleed, superior inertness and consistent reproducibility.
Manufacturer:  Thermo Scientific™ 260755760
 View more versions of this product
Catalog No. 26-075-5760
  Check Availability
Add to cart
Specifications
Description
Analyze organophosphorus pesticides with ultralow bleed, superior inertness, and consistent reproducibility using Thermo Scientific™ TraceGOLD™ TG-OPP I and II GC Columns.This column's low bleed delivers excellent signal-to-noise ratio, sensitivity, and mass spectral integrity. Expect full separation of organophosphorus pesticides with a fast analysis time. TraceGOLD TG-OPP I and II GC Columns are ideal for U.S. EPA methods 8141A.
Description & Specifications
Specifications
GC/MS
0.32mm
TraceGold TG-OPP II
330°C
30m