Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TraceGOLD™ TG-WaxMS GC Columns

Analyze FAMEs with excellent thermal stability using this column.

$854.00 - $1722.00

Specifications

Stationary Phase TraceGOLD™ TG-WAXMS
USP Type G14, G15, G16, G20, G39
Max. Temperature 240/260°C
Polarity Polar
View More Specs

Products 16
Catalog Number Mfr. No. Diameter (Metric) Film Thickness Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Film Thickness Length (Metric) Price Quantity & Availability  
26-088-1420
Accessories View Documents
Thermo Scientific™
260881420
0.25 mm 0.25 μm 30 m
Each for $854.00
Add to Cart
 
26-088-1430
Accessories View Documents
Thermo Scientific™
260881430
0.32 mm 0.25 μm 30 m
Each for $1,131.00
Add to Cart
 
26-088-1440
Accessories View Documents
Thermo Scientific™
260881440
0.53 mm 0.25 μm 30 m
Each for $1,026.00
Add to Cart
 
26-088-1550
Accessories View Documents
Thermo Scientific™
260881550
0.32 mm 0.25 μm 60 m
Each for $1,606.00
Add to Cart
 
26-088-1560
Accessories View Documents
Thermo Scientific™
260881560
0.53 mm 0.25 μm 60 m
Each for $1,722.00
Add to Cart
 
26-088-2240
Accessories View Documents
Thermo Scientific™
260882240
0.32 mm 0.5 μm 30 m
Each for $1,106.00
Add to Cart
 
26-088-2250
View Documents
Thermo Scientific™
260882250
0.53 mm 0.5 μm 30 m
Each for $1,278.00
Add to Cart
 
26-088-2350
Accessories View Documents
Thermo Scientific™
260882350
0.25 mm 0.5 μm 60 m
Each for $1,529.00
Add to Cart
 
26-088-2360
Accessories View Documents
Thermo Scientific™
260882360
0.32 mm 0.5 μm 60 m
Each for $1,588.00
Add to Cart
 
26-088-2370
Accessories View Documents
Thermo Scientific™
260882370
0.53 mm 0.5 μm 60 m
Each for $1,722.00
Add to Cart
 
26-088-2970
View Documents
Thermo Scientific™
260882970
0.32 mm 1 μm 30 m
Each for $1,106.00
Add to Cart
 
26-088-2980
Accessories View Documents
Thermo Scientific™
260882980
0.53 mm 1 μm 30 m
Each for $1,271.00
Add to Cart
 
26-088-3090
Accessories View Documents
Thermo Scientific™
260883090
0.32 mm 1 μm 60 m
Each for $1,641.00
Add to Cart
 
26-088-3100
Accessories View Documents
Thermo Scientific™
260883100
0.53 mm 1 μm 60 m
Each for $1,642.00
Add to Cart
 
26-088-3360
Accessories View Documents
Thermo Scientific™
260883360
0.53 mm 1.5 μm 30 m
Each for $1,055.00
Add to Cart
 
26-088-3370
View Documents
Thermo Scientific™
260883370
0.53 mm 1.5 µm 60 m Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
Description

Description

Analyze FAMEs with excellent thermal stability using the Thermo ScientificTM TraceGOLDTM TG-WaxMS GC Column.

Engineered for Superior Performance
  • Polar phase, polyethylene glycol
  • Polar-deactivated surface that tightly binds polymer for excellent thermal stability
  • Resistive to oxidative damage and damage from strongly acidic or basic volatiles better than silicone solid phases
  • Applications:
  • Fatty acid methyl esters (FAMEs)
  • Flavor compounds and essential oils
  • Solvents
  • Xylene isomers
  • EPA Method 603 for Acrolein/Acrylonitrile
A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • This low-bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Recommended For:
  • Aldehydes
  • Essential oils
  • Solvents
  • Xylenes
  • Alternative Uses:
  • Acids
  • Alcohols
  • Alcohols in beverages
  • Alditol acetates (sugars)
  • Amines–Aliphatic
  • Amines–Aromatic
  • Herbicides
  • Ketones
  • Nitroaromatics
  • Phenols
  • Volatiles
Similar To:
  • DB-WAX
  • DB-WAXetr
  • HP-Wax
  • HP-Innowax
  • SUPELCOWAX™ 10
  • CP-Wax 52 CB
  • Stabilwax™
  • Rtx™-Wax
  • BP20
  • ZB-Wax
  • Optima™ Wax
  • AT™-Wax
Specifications

Specifications

TraceGOLD™ TG-WAXMS
G14, G15, G16, G20, G39
240/260°C
Polar
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit