Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRACP/PAP/ACP5 Antibody (rACP5/1070), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275780AF647
Description
TRACP/PAP/ACP5 Monoclonal specifically detects TRACP/PAP/ACP5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TRACP/PAP/ACP5 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ACP5 | |
Recombinant full-length human TRACP/PAP/ACP5 protein (Uniprot: P13686) | |
0.1 mL | |
Protein Phosphatase | |
It recognizes a protein of 35kDa, which is identified as tartrate-resistant acid phosphatase (TRAcP). It exists as two isoforms (5a and 5b). This monoclonal antibody reacts with both the isoforms. Serum TRAcP 5a is secreted by macrophages and dendritic cells and increased in many patients of rheumatoid arthritis.Serum TRAcP 5b is produced from osteoclasts and elevated during bone resorption. TRAcP is an iron containing glycoprotein, which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is the most basic of the acid phosphatases and is the only form not inhibited by L(+)-tartrate. TRAcP is synthesized as a latent proenzyme and is activated by proteolytic cleavage and reduction. Normally, TRAcP is highly expressed by osteoclasts, activated macrophages, neurons and endometrium during pregnancy. Expression of TRAcP is increased in certain pathological conditions such as Leukemic Reticuloendotheliosis (Hairy Cell Leukemia), Gauchers Disease, HIV-induc | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
rACP5/1070 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG1 κ |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction