Learn More
Aviva Systems Biology Corp TRADD Antibody - middle region (AVARP02012_P050) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP02012_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRADD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRADD. Alternate names: Hs.89862, MGC11078ApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-62476