Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products Medium Duty Synthetic Leather Mechanics Gloves

Mesh fabric back gloves offer comfort and breathability, while synthetic leather patches provide additional protection.

$11.50

Specifications

Color Black
For Use With (Application) Mechanics, Construction, Fabrication, and Assembly
Glove Cuff Style Hook and Loop
Back Material Mesh
Material Synthetic Leather
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-555-039
View Documents
Pyramex Safety Products
GL102S
Small
Each for $11.50
Add to cart
 
19-555-038
View Documents
Pyramex Safety Products
GL102M
Medium
Each for $11.50
Add to cart
 
19-555-037
View Documents
Pyramex Safety Products
GL102L
Large
Each for $11.50
Add to cart
 
19-555-041
View Documents
Pyramex Safety Products
GL102XL
X-Large
Each for $11.50
Add to cart
 
19-555-040
View Documents
Pyramex Safety Products
GL102X2
2X-Large
Each for $11.50
Add to cart
 
Description

Description

  • Double layer spandex with mesh for comfort and breathability
  • Silicone palm patch for anti-slip and enhanced grip capabilities
  • Thicker embossed neoprene on knuckles for additional protection
  • Touch screen function on 2 fingers and thumb
Specifications

Specifications

Black
Hook and Loop
Synthetic Leather
GL102
Mechanics, Construction, Fabrication, and Assembly
Mesh
Mechanics Gloves
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit