Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-1, Mouse anti-Human, DyLight 755, Clone: TRAF1/2770, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279924IR
Description
TRAF-1 Monoclonal antibody specifically detects TRAF-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
TRAF-1 | |
TRAF1/2770 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
TRAF1 | |
Recombinant fragment of human TRAF-1 protein (around aa 73-219) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1 | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Apoptosis, Cancer, Cytokine Research | |
Primary | |
This monoclonal antibody recognizes a protein of 52kDa, which is identified as TRAF-1 (TNFR-associated factor 1), which is highly useful for the differentiation of LyP and cALCL in patients with cutaneous CD30-positive lymphoproliferations. CD30-positive lymphoproliferations of the skin comprise 30% of all primary cutaneous T-cell lymphomas (CTCLs). Besides borderline cases this group includes lymphomatoid papulosis (LyP) and primary cutaneous anaplastic large T-cell lymphoma (cALCL). |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title