Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-1, Mouse anti-Human, Janelia Fluor 646, Clone: TRAF1/2770, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279924JF646

Catalog No. NB012685


Add to cart

Description

Description

TRAF-1 Monoclonal antibody specifically detects TRAF-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TRAF-1
TRAF1/2770
Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TRAF1
Recombinant fragment of human TRAF-1 protein (around aa 73-219) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1
Mouse
IgG1 Kappa
0.1 mL
Apoptosis, Cancer, Cytokine Research
Primary
This monoclonal antibody recognizes a protein of 52kDa, which is identified as TRAF-1 (TNFR-associated factor 1), which is highly useful for the differentiation of LyP and cALCL in patients with cutaneous CD30-positive lymphoproliferations. CD30-positive lymphoproliferations of the skin comprise 30% of all primary cutaneous T-cell lymphomas (CTCLs). Besides borderline cases this group includes lymphomatoid papulosis (LyP) and primary cutaneous anaplastic large T-cell lymphoma (cALCL).
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit