Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen TRAF-2
Applications Immunofluorescence, Immunoprecipitation, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092547
View Documents
Novus Biologicals
NBP303438100UL
100 μg Each for $320.71
Add to cart
 
NB092548
View Documents
Novus Biologicals
NBP30343820UL
20 μg Each for $138.37
Add to cart
 
Description

Description

TRAF-2 Polyclonal antibody specifically detects TRAF-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown
Specifications

Specifications

TRAF-2
Unconjugated
Rabbit
Apoptosis, Cancer, Cytokine Research, Zinc Finger
PBS with 50% glycerol, pH7.3.
7186
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Immunoprecipitation, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TRAF-2 (NP_066961.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit