Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB10056177SS
Description
TRAF-4 Polyclonal specifically detects TRAF-4 in Human, Mouse, Rat, Bovine, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
TRAF-4 | |
Polyclonal | |
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:1000-1:5000, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
CART1RING finger protein 83, Cysteine-rich domain associated with RING and Traf domains protein 1, Metastatic lymph node gene 62 protein, MLN 62, MLN62cysteine-rich domain associated with ring and TRAF domain, RNF83malignant 62, TNF receptor-associated factor 4, tumor necrosis receptor-associated factor 4A | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
TRAF4 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antisera with 0.02% Sodium Azide | |
TRAF4 | |
A synthetic peptide corresponding to amino acids 1-17 (MPGFDYKFLEKPKRRLL) of human TRAF4 was used as immunogen; GenBank no. gi 22027622 ref NP_004286.2 . | |
0.025 mL | |
Cancer, Zinc Finger | |
9618 | |
Human, Mouse, Rat, Bovine, Zebrafish | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction