Learn More
TRAF3IP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549934
Catalog No. PA549934
Add to cart
Description
Description
The antibody detects endogenous levels of total TRAF3IP3 / T3JAM protein.
The gene encodes a protein that mediates cell growth by modulating the c-Jun N-terminal kinase signal transduction pathway. The encoded protein may also interact with a large multi-protein assembly containing the phosphatase 2A catalytic subunit. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2013].
Specifications
Specifications
TRAF3IP3
Polyclonal
Unconjugated
TRAF3IP3
Liquid
TRAF3IP3
Synthesized peptide derived from internal of human T3JAM.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9Y228, Q8C0G2
T3JAM
Rabbit
IgG
100 μL
-20°C
Primary
215243, 360900, 80342