Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF5 Mouse anti-Human, Clone: 55A219, Abnova™

Mouse monoclonal antibody raised against human TRAF5.

Manufacturer:  Abnova Corporation MAB12540

Catalog No. 89-935-049


Add to cart

Description

Description

The scaffold protein encoded by this gene is a member of the tumor necrosis factor receptor-associated factor (TRAF) protein family and contains a meprin and TRAF homology (MATH) domain, a RING-type zinc finger, and two TRAF-type zinc fingers. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. This protein is one of the components of a multiple protein complex which binds to tumor necrosis factor (TNF) receptor cytoplasmic domains and mediates TNF-induced activation. Alternate transcriptional splice variants have been characterized. [provided by RefSeq]

Specifications

Specifications

TRAF5
55A219
Mouse monoclonal antibody raised against human TRAF5.
In PBS (0.05% BSA, 0.05% sodium azide).
MGC:39780/RNF84
Mouse
IgG
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TRAF5
Recombinant fusion protein corresponding to amino acids 77-186 of human TRAF5.
Protein G purification
RUO
Primary
7188
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit