Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF7 Rabbit anti-Human, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Manufacturer:  Abnova Corporation PAB30536

Catalog No. 89-032-878


Add to cart

Description

Description

Rabbit polyclonal antibody raised against recombinant human TRAF7.

  • High specificity
  • High reactivity
Specifications

Specifications

TRAF7
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200), Immunofluorescence (1-4 ug/mL), The optimal working dilution should be determined by the end user.
Q6Q0C0
DKFZp586I021|MGC7807|RFWD1|RNF119
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against recombinant human TRAF7.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Liquid
TRAF7
Recombinant protein corresponding to human TRAF7.
Antigen affinity purification
RUO
Primary
84231
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)