Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAFD1 Mouse anti-Human, Allophycocyanin, Clone: 8E6E7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237408APC

Catalog No. NBP237408T

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TRAFD1
8E6E7
Flow Cytometry
Purified
TRAFD1
Purified recombinant fragment of human TRAFD1 (AA: 401-582) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
FLN29FLN29 gene product, Protein FLN29, TRAF-type zinc finger domain containing 1, TRAF-type zinc finger domain-containing protein 1
Mouse
IgG1
0.1 ml
Primary
10906
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit