Learn More
TRAFD1 Mouse anti-Human, DyLight 550, Clone: 8E6E7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237408R
Catalog No. NBP237408R
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
TRAFD1
8E6E7
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Purified
TRAFD1
Purified recombinant fragment of human TRAFD1 (AA: 401-582) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
FLN29FLN29 gene product, Protein FLN29, TRAF-type zinc finger domain containing 1, TRAF-type zinc finger domain-containing protein 1
Mouse
IgG1
0.1 ml
Primary
10906