Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (32A1380), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056747PECY55
Description
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Flow Cytometry.Specifications
TRAILR1/TNFRSF10A | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
TNFRSF10A | |
This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4. | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
32A1380 | |
Flow Cytometry | |
APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
Mouse | |
Protein G purified | |
Apoptosis, Cancer | |
8797.0 | |
Store at 4°C in the dark. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction