Learn More
TRAIL-R1 (CD261) Mouse anti-Human, BUV395, Clone: S35-934, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 745583
Catalog No. BDB745583
Add to cart
Description
Description
TRAIL-R1 (CD261) Monoclonal antibody specifically detects TRAIL-R1 (CD261) in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TRAIL-R1 (CD261)
S35-934
Brilliant Ultraviolet 395
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Human