Learn More
TRAIL-R2 (CD262) Mouse anti-Human, PE, Clone: YM366, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565499
Catalog No. BDB565499
Add to cart
Description
Description
TRAIL-R2 (CD262) Monoclonal antibody specifically detects TRAIL-R2 (CD262) in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
TRAIL-R2 (CD262)
YM366
Vol. Per test: 5μL
Purified
Murine
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions
Monoclonal
Flow Cytometry
PE
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
TNFRSF10B, DR5, KILLER, TRAILR2, TRICK2, ZTNFR9
Human DR5/TRAIL-R2 Recombinant Protein
100 Tests
Cell Biology
Primary
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.