Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAIL/TNFSF10 Mouse anti-Human, DyLight 755, Clone: 2E5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP140925IR

Catalog No. NBP140925IR


Add to cart

Description

Description

TRAIL/TNFSF10 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Western Blotting.
Specifications

Specifications

TRAIL/TNFSF10
200000
Western Blot, Flow Cytometry
Purified
TNFSF10
Recombinant human CD253 (TRAIL) aa 95-281.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody recognises the 281 amino acid protein termed TNF related apoptosis-inducing ligand (TRAIL), also known as CD253.
Flow Cytometry, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
Mouse
IgG1
0.25 ml
Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Signal Transduction, Tumor Suppressors
Primary
8743
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit