Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (32A242) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 8 publications
$206.50 - $499.50
Specifications
Antigen | TRAILR1/TNFRSF10A |
---|---|
Clone | 32A242 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 - 3 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23988408), Immunoblotting reported in scientific literature (PMID 31534206) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056528T
![]() |
Novus Biologicals
NB10056528SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056528
![]() |
Novus Biologicals
NB10056528 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoblotting.Specifications
TRAILR1/TNFRSF10A | |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer | |
O00220 | |
8797 | |
This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
32A242 | |
Western Blot 1 - 3 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23988408), Immunoblotting reported in scientific literature (PMID 31534206) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
TNFRSF10A | |
IgG1 | |
Protein G purified | |
In HeLa, a 57 kDa band should be observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title