Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TRAILR1/TNFRSF10A Antibody Pair [HRP]

Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795995PLATES

 View more versions of this product

Catalog No. NB010238


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TRAILR1/TNFRSF10A.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
2.34 pg/mL
8797
RUO
TRAILR1/TNFRSF10A
Apoptosis, Cancer
Human
TNFRSF10A
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 150 pg/mL is recommended.
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TRAILR1/TNFRSF10A.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human TRAILR1/TNFRSF10A monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of mouse anti-Human TRAILR1/TNFRSF10A monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 30 ng of recombinant Human TRAILR1/TNFRSF10A.
  • Storage is content dependent.
    Rabbit/Mouse
    5 Plates
    2.34-150 pg/mL
    APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit