Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR2/TNFRSF10B Antibody (JA03-38), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP267225

Catalog No. NBP267225


Add to Cart

Description

Description

TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TRAILR2/TNFRSF10B
Monoclonal
1 mg/mL
Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Rabbit
Protein A purified
Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway
8795
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
JA03-38
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
TNFRSF10B
Synthetic peptide within Human TRAILR2/TNFRSF10B aa 391-440/ 440. (SwissProt: O14763 Human)
100 μL
Primary
Human
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit