Learn More
Human TRAILR4/TNFRSF10D Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB633RG100UG
Catalog No. FAB633RG100
Add to Cart
This item is not returnable. View return policy
Description
Description
TRAILR4/TNFRSF10D/DcR2 Monoclonal antibody specifically detects TRAILR4/TNFRSF10D/DcR2 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
TRAILR4/TNFRSF10D/DcR2
Monoclonal
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TNFRSF10D, TRAIL R4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain
Mouse myeloma cell line NS0-derived recombinant human TRAIL R4/TNFRSF10D, Ala56-His211, Accession # Q9UBN6
100 μg
Primary
Human
Purified
Flow Cytometry
104918R
Flow Cytometry 0.25-1 ug/106 cells
Q9UBN6
Mouse
Protein A or G purified from cell culture supernatant
RUO
8793
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG1
Product Suggestions
Product Suggestions