Tocris™ TRAM 39

Potent KCa3.1 blocker

Manufacturer: Tocris™ 4952/50

 View more versions of this product

Catalog No. 49-525-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors TRAM 39
    Purity >99
    Molecular Weight 303.78
    Quantity 50mg
    Product Type TRAM 39

    Potent intermediate conductance Ca2+-activated K+ channel (KCa3.1) blocker (Kd = 60nM). Has no effect on cytochrome p450 activity. Inhibits I-EBIO-stimulated increases in rat artery membrane potential ex vivo. Also diminishes LPS-induced cryptidin (mammalian α-defensin) release from paneth cells in vitro.

    • Chemical Name: 2-Chloro-α,α-diphenylbenzeneacetonitrile
    • Formula: C20H14ClN
    • CAS Number: 197525-99-8
    • Solubility: Soluble to 50mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit