Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tranilast, JP, 99-101%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
53902-12-8
C18H17NO5
327.34
NZHGWWWHIYHZNX-CSKARUKUSA-N
2-[(2E)-3-(3,4-dimethoxyphenyl)prop-2-enamido]benzoic acid
COC1=CC=C(\C=C\C(=O)NC2=CC=CC=C2C(O)=O)C=C1OC
53902-12-8
C18H17NO5
327.34
NZHGWWWHIYHZNX-CSKARUKUSA-N
2-[(2E)-3-(3,4-dimethoxyphenyl)prop-2-enamido]benzoic acid
COC1=CC=C(\C=C\C(=O)NC2=CC=CC=C2C(O)=O)C=C1OC

T1698, 53902-12-8, C18H17NO5

Spectrum™ Tranilast, JP is an anti-allergy drug used against asthma, autoimmune diseases, and atopic and fibrotic pathologies. All Spectrum Chemical JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Ignition Residue 0.002
Loss on Drying 0.003
Quantity 10 kg
Melting Point 207° to 210°C
CAS Max % 1
Percent Purity 99 to 101%