Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Bis(tri-n-butylstannyl)ethylene, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
14275-61-7
C26H56Sn2
606.15
MFCD01631299
VNKOWRBFAJTPLS-UHFFFAOYSA-N
trans-1,2-bis tributylstannyl ethene,e-1,2-bis tributylstannyl ethene,trans-1,2-bis tri-n-butylstannyl ethylene,tributyl e-2-tributylstannyl ethenyl stannane,stannane, 1e-1,2-ethenediylbis tributyl,stannane, 1,2-ethenediylbis tributyl,e-1,2-bis-tri-n-butylstannyl-ethylene,tributyl 2-tributylstannyl ethenyl stannane
5378370
tributyl-[(E)-2-tributylstannylethenyl]stannane
CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC
14275-61-7
C26H56Sn2
606.15
MFCD01631299
VNKOWRBFAJTPLS-UHFFFAOYSA-N
trans-1,2-bis tributylstannyl ethene,e-1,2-bis tributylstannyl ethene,trans-1,2-bis tri-n-butylstannyl ethylene,tributyl e-2-tributylstannyl ethenyl stannane,stannane, 1e-1,2-ethenediylbis tributyl,stannane, 1,2-ethenediylbis tributyl,e-1,2-bis-tri-n-butylstannyl-ethylene,tributyl 2-tributylstannyl ethenyl stannane
5378370
tributyl-[(E)-2-tributylstannylethenyl]stannane
CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC
Refractive Index 1.5000 to 1.5030 (20°C, 589nm)
Quantity 1g
Color Colorless to Yellow
Infrared Spectrum Authentic
NMR 94% min.
Packaging Glass bottle
Assay Percent Range 95%
Linear Formula (CH3(CH2)3)3SnCHCHSn((CH2)3CH3)3
Formula Weight 606.15
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material trans-1, 2-Bis(tri-n-butylstannyl)ethylene
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger