Learn More
trans-1,2-Bis(tri-n-butylstannyl)ethylene, 95%, ACROS Organics™
Specifications
NMR | 94% min. |
Packaging | Glass bottle |
Refractive Index | 1.5000 to 1.5030 (20°C, 589nm) |
Quantity | 5g |
Color | Colorless to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Linear Formula | (CH3(CH2)3)3SnCHCHSn((CH2)3CH3)3 |
Formula Weight | 606.15 |
Physical Form | Liquid |
Show More |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger