Alfa Aesar™ trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, 98%

Manufacturer: Alfa Aesar™ B2292814

Quantity 25g

 View more versions of this product

Catalog No. AAB2292814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 125572-95-4
    Molecular Formula C14H22N2O8·H2O
    Formula Weight 364.35 (346.34 Anhydrous)
    MDL Number MFCD00149243
    Beilstein 2226756
    Synonym (1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate; CyDTA
    Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

    Specifications

    Formula Weight 364.35 (346.34 Anhydrous)
    Synonym (1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate; CyDTA
    Melting Point ≈220°C (dec.)
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C14H22N2O8·H2O
    MDL Number MFCD00149243
    CAS 125572-95-4
    Beilstein 2226756
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
    Assay 98%

    Description

    Applications
    trans-1,2-Diaminocyclohexane-N,N,N ft.,N ft.-tetraacetic acid monohydrate is an analytical reagent and a complexating agent for sodium. It acts as a ligand to prepare lanthanide shift reagents. It is involved in the separation and determination of iron(III)-dimethyldithiocarbamate, manganese(II)-ethylenebisdithiocarbamate and zinc(II)-ethylenebisdithiocarbamate by sensitive capillary electrophoretic ultraviolet method.

    Solubility
    Slightly soluble in water. Soluble in 1N sodium hydroxide and alkali solutions. Insoluble in most common organic solvents.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit