Learn More
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, ACS, 97.5%-100.5%, Thermo Scientific Chemicals
Description
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate is an analytical reagent and a complexating agent for sodium. It acts as a ligand to prepare lanthanide shift reagents. It is involved in the separation and determination of iron(III)-dimethyldithiocarbamate, manganese(II)-ethylenebisdithiocarbamate and zinc(II)-ethylenebisdithiocarbamate by sensitive capillary electrophoretic ultraviolet method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 213°C to 216°C |
Assay Percent Range | 97.5 to 100.5% |
Quantity | 5 g |
Beilstein | 3222748 |
Solubility Information | Slightly soluble in water. Soluble in 1N sodium hydroxide and alkali solutions. Insoluble in most common organic solvents. |
Formula Weight | 364.35 (346.34 Anhydrous) |
Percent Purity | 97.5 to 100.5% |
Grade | ACS |
Physical Form | Solid |
Chemical Name or Material | trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, For analysis ACS |
Safety and Handling
Hazard Category | H319 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P264b-P280i-P305+P351+P338 |
EINECSNumber | 236-308-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.