Alfa Aesar™ trans-1-Fluoro-4-(4-n-heptylcyclohexyl)benzene, 97%

Manufacturer: Alfa Aesar™ H5527406

Quantity 5g

 View more versions of this product

Catalog No. AAH5527406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76802-59-0
    Molecular Formula C19H29F
    Formula Weight 276.44
    MDL Number MFCD11053424
    Chemical Name or Material trans-1-Fluoro-4-(4-n-heptylcyclohexyl)benzene

    Specifications

    Formula Weight 276.44
    Molecular Formula C19H29F
    MDL Number MFCD11053424
    CAS 76802-59-0
    Melting Point 35° to 37°C
    Quantity 5g
    Chemical Name or Material trans-1-Fluoro-4-(4-n-heptylcyclohexyl)benzene
    Assay 97%

    Description

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit