Alfa Aesar™ trans-1-Hepten-1-ylboronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H2787606

Quantity 5g

 View more versions of this product

Catalog No. AAH2787606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 169339-75-7
    Molecular Formula C13H25BO2
    Formula Weight 224.15
    MDL Number MFCD05663884
    Synonym E-2-(1-Heptenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Chemical Name or Material trans-1-Hepten-1-ylboronic acid pinacol ester

    Specifications

    Formula Weight 224.15
    Molecular Formula C13H25BO2
    MDL Number MFCD05663884
    CAS 169339-75-7
    Synonym E-2-(1-Heptenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Boiling Point 73° to 77°C (0.5mmHg)
    Quantity 5g
    Flash Point 106°C (223°F)
    Chemical Name or Material trans-1-Hepten-1-ylboronic acid pinacol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit