Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
70708-33-7
C8H18N2·2ClH
215.17
UDYXZFBJGBDHHP-RHJRFJOKSA-N
1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride,trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride,1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans,trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride,trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride,1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride,1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride
44890801
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride
CNC1CCCCC1NC.Cl.Cl
70708-33-7
C8H18N2·2ClH
215.17
UDYXZFBJGBDHHP-RHJRFJOKSA-N
1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride,trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride,1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans,trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride,trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride,1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride,1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride
44890801
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride
CNC1CCCCC1NC.Cl.Cl
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 97.5 to max. 102.5 % min. (Argentometry)
Formula Weight 215.17
Percent Purity 98%
Chemical Name or Material trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist

GHS Signal Word: Warning