Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile Matrix Substance for MALDI-MS, ≥99.0%, MilliporeSigma™ Supelco™
Specifications
Packaging | Glass Bottle |
---|---|
CAS | 300364-84-5 |
Type | Matrix Substance for MALDI-MS |
Linear Formula | C17H18N2 |
MDL Number | MFCD03453260 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-3852
|
MilliporeSigma Supelco
87884250MGF |
250 mg | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
11-101-3851
|
MilliporeSigma Supelco
878841GF |
1 g | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Specifications
Glass Bottle | |
Matrix Substance for MALDI-MS | |
MFCD03453260 | |
250.34 | |
Matrix Substance for MALDI-MS |
300364-84-5 | |
C17H18N2 | |
DCTB | |
≥99.0% (HPLC) | |
123°C to 126°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title