Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2,3,4-Trimethoxycinnamic acid, 99%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
33130-03-9
C12H14O5
238.239
MFCD00014376
ZYOPDNLIHHFGEC-FNORWQNLSA-N
2,3,4-trimethoxycinnamic acid,trans-2,3,4-trimethoxycinnamic acid,3-2,3,4-trimethoxyphenyl acrylic acid,2e-3-2,3,4-trimethoxyphenyl prop-2-enoic acid,2-propenoic acid, 3-2,3,4-trimethoxyphenyl,3-2,3,4-trimethoxyphenyl propenoic acid,trans-2, 3, 4-trimethoxycinnamate,2e-3-2,3,4-trimethoxyphenyl-2-propenoic acid,e-3-2,3,4-trimethoxyphenyl acrylic acid,2e-3-2,3,4-trimethoxyphenyl acrylic acid
735841
(E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoic acid
COC1=C(C(=C(C=C1)C=CC(=O)O)OC)OC
33130-03-9
C12H14O5
238.239
MFCD00014376
ZYOPDNLIHHFGEC-FNORWQNLSA-N
2,3,4-trimethoxycinnamic acid,trans-2,3,4-trimethoxycinnamic acid,3-2,3,4-trimethoxyphenyl acrylic acid,2e-3-2,3,4-trimethoxyphenyl prop-2-enoic acid,2-propenoic acid, 3-2,3,4-trimethoxyphenyl,3-2,3,4-trimethoxyphenyl propenoic acid,trans-2, 3, 4-trimethoxycinnamate,2e-3-2,3,4-trimethoxyphenyl-2-propenoic acid,e-3-2,3,4-trimethoxyphenyl acrylic acid,2e-3-2,3,4-trimethoxyphenyl acrylic acid
735841
(E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoic acid
COC1=C(C(=C(C=C1)C=CC(=O)O)OC)OC
Quantity 5g
Color Yellow
Melting Point 172°C to 174°C
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula (CH3O)3C6H2CH=CHCOOH
Beilstein 10, III, 2197
Formula Weight 238.24
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material trans-2, 3, 4-Trimethoxycinnamic acid, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.