Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-2,3-Epoxybutane, 97%

Click to view available options
Quantity:
1g
5g
25g
21490-63-1
C4H8O
72.107
MFCD00005129
PQXKWPLDPFFDJP-QWWZWVQMSA-N
trans-2,3-dimethyloxirane,trans-2,3-epoxybutane,2r,3r-2,3-dimethyloxirane,oxirane, 2,3-dimethyl-, trans,trans-2-butene oxide,butane, 2,3-epoxy-, trans,e-2r,3r-epoxybutane,trans-,a-butylene oxide,trans-2r,3r-epoxybutane,2,3-dimethyloxirane, trans
6432237
(2R,3R)-2,3-dimethyloxirane
CC1C(O1)C
21490-63-1
C4H8O
72.107
MFCD00005129
PQXKWPLDPFFDJP-QWWZWVQMSA-N
trans-2,3-dimethyloxirane,trans-2,3-epoxybutane,2r,3r-2,3-dimethyloxirane,oxirane, 2,3-dimethyl-, trans,trans-2-butene oxide,butane, 2,3-epoxy-, trans,e-2r,3r-epoxybutane,trans-,a-butylene oxide,trans-2r,3r-epoxybutane,2,3-dimethyloxirane, trans
6432237
(2R,3R)-2,3-dimethyloxirane
CC1C(O1)C
Applications
A higher order cuprate reacted with trans-2,3-epoxybutane a high yield both counted on the epoxide (96%) and on the haloalkane (85%). Enolate derived from trans-2,3-Epoxybutan has chiral recognition.

Solubility
Soluble in water(95g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Quantity 5g
Boiling Point 54°C to 55°C
Odor Recognizable
Flash Point −26°C (−14°F)
UN Number UN3271
Beilstein 79772
Formula Weight 72.11
Percent Purity 97%
Chemical Name or Material trans-2,3-Epoxybutane

GHS H Statement
H225-H341-H315-H319-H335
Highly flammable liquid and vapour.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

RTECSNumber : EK3855040