Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-2,4-Difluorocinnamic acid, 97%

Click to view available options
Quantity:
5g
25g
94977-52-3
C9H6F2O2
184.142
MFCD00010317
PQDXPFJQTKGTFP-DUXPYHPUSA-N
2,4-difluorocinnamic acid,e-3-2,4-difluorophenyl acrylic acid,trans-2,4-difluorocinnamic acid,3-2,4-difluorophenyl acrylic acid,2e-3-2,4-difluorophenyl prop-2-enoic acid,2,4-difluorocinnamicacid,3-2,4-difluorophenyl-2-propenoic acid,2e-3-2,4-difluorophenyl acrylic acid,rarechem bk hw 0121,e-3-2,4-difluorophenyl prop-2-enoic acid
5374945
(E)-3-(2,4-difluorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1F)F)C=CC(=O)O
94977-52-3
C9H6F2O2
184.142
MFCD00010317
PQDXPFJQTKGTFP-DUXPYHPUSA-N
2,4-difluorocinnamic acid,e-3-2,4-difluorophenyl acrylic acid,trans-2,4-difluorocinnamic acid,3-2,4-difluorophenyl acrylic acid,2e-3-2,4-difluorophenyl prop-2-enoic acid,2,4-difluorocinnamicacid,3-2,4-difluorophenyl-2-propenoic acid,2e-3-2,4-difluorophenyl acrylic acid,rarechem bk hw 0121,e-3-2,4-difluorophenyl prop-2-enoic acid
5374945
(E)-3-(2,4-difluorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1F)F)C=CC(=O)O
Melting Point 215°C to 219°C
Quantity 5g
Beilstein 3129944
Formula Weight 184.14
Percent Purity 97%
Chemical Name or Material trans-2,4-Difluorocinnamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures