Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (±)-trans-2,5-Bis-(3,4,5-trimethoxyphenyl)-1,3-dioxolane

Click to view available options
Quantity:
1mg
5mg
116673-45-1
C21H26O8
406.431
MFCD00155367
DUAYHYGJMLZLCE-NQIIRXRSSA-N
+/-trans-2,5-bis 3,4,5-trimethoxyphenyl-1,3-dioxolane,bis-3,4,5-trimethoxyphenyl-1,3-dioxolane,2s,4s-2,4-bis 3,4,5-trimethoxyphenyl-1,3-dioxolane
53779199
(2S,4S)-2,4-bis(3,4,5-trimethoxyphenyl)-1,3-dioxolane
COC1=CC(=CC(=C1OC)OC)C2COC(O2)C3=CC(=C(C(=C3)OC)OC)OC
116673-45-1
C21H26O8
406.431
MFCD00155367
DUAYHYGJMLZLCE-NQIIRXRSSA-N
+/-trans-2,5-bis 3,4,5-trimethoxyphenyl-1,3-dioxolane,bis-3,4,5-trimethoxyphenyl-1,3-dioxolane,2s,4s-2,4-bis 3,4,5-trimethoxyphenyl-1,3-dioxolane
53779199
(2S,4S)-2,4-bis(3,4,5-trimethoxyphenyl)-1,3-dioxolane
COC1=CC(=CC(=C1OC)OC)C2COC(O2)C3=CC(=C(C(=C3)OC)OC)OC
Quantity 1mg
Color White
Formula Weight 406.43
Physical Form Solid
Chemical Name or Material (±)-trans-2,5-Bis-(3,4,5-trimethoxyphenyl)-1,3-dioxolane