Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-2,5-Dimethylpiperazine, 98%

Click to view available options
Quantity:
5g
25g
100g
2815-34-1
C6H14N2
114.192
MFCD00070516
NSMWYRLQHIXVAP-OLQVQODUSA-N
trans-2,5-dimethylpiperazine,2r,5s-2,5-dimethylpiperazine,2s,5r-2,5-dimethylpiperazine,2s,5r-2,5-dimethyl-piperazine,piperazine, 2,5-dimethyl-, 2r,5s-rel,2r,5s-2,5-dimethylpiperazin,piperazine, 2,5-dimethyl-, trans,2r,5s-2,5-dimethyl-piperazine,pubchem15878,2,5-dimethylpiperazine #
220672
(2R,5S)-2,5-dimethylpiperazine
CC1CNC(CN1)C
2815-34-1
C6H14N2
114.192
MFCD00070516
NSMWYRLQHIXVAP-OLQVQODUSA-N
trans-2,5-dimethylpiperazine,2r,5s-2,5-dimethylpiperazine,2s,5r-2,5-dimethylpiperazine,2s,5r-2,5-dimethyl-piperazine,piperazine, 2,5-dimethyl-, 2r,5s-rel,2r,5s-2,5-dimethylpiperazin,piperazine, 2,5-dimethyl-, trans,2r,5s-2,5-dimethyl-piperazine,pubchem15878,2,5-dimethylpiperazine #
220672
(2R,5S)-2,5-dimethylpiperazine
CC1CNC(CN1)C
Flash Point 58°C (136°F)
Quantity 25g
Boiling Point 162°C to 165°C
Melting Point 115°C to 119°C
UN Number UN2925
Beilstein 79886
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material trans-2,5-Dimethylpiperazine

GHS H Statement
H314-H318-H228-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Harmful in contact with skin.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

RTECSNumber : TL5950000