Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hexene, 99%, Thermo Scientific Chemicals

Catalog Number AAB2228909
Click to view available options
Quantity:
2.5 g
10 g
4050-45-7
C6H12
84.16
MFCD00009473
RYPKRALMXUUNKS-HWKANZROSA-N
trans-2-hexene, e-2-hexene, 2-hexene, e, hexene, isomer, hexene, isomers, unii-2tz30ggg1a, 2e-hexene, e-hex-2-ene, 2tz30ggg1a, 2-hexene, 2e
639661
CCC\C=C\C
4050-45-7
C6H12
84.16
MFCD00009473
RYPKRALMXUUNKS-HWKANZROSA-N
trans-2-hexene, e-2-hexene, 2-hexene, e, hexene, isomer, hexene, isomers, unii-2tz30ggg1a, 2e-hexene, e-hex-2-ene, 2tz30ggg1a, 2-hexene, 2e
639661
CCC\C=C\C

trans-2-Hexene is used as a co monomer in the production of polyethylene. The use of trans-2-hexene, which is derived from 3-hexyne using sodium in liquid ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Hexene is used as a co monomer in the production of polyethylene. The use of trans-2-hexene, which is derived from 3-hexyne using sodium in liquid ammonia.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point -99°C
Density 0.674
Boiling Point 68°C to 69°C
Flash Point −20°C (−4°F)
Odor Mild
Refractive Index 1.394
Quantity 10 g
UN Number UN3295
Beilstein 1719020
Solubility Information Difficult to mix.
IUPAC Name (E)-hex-2-ene
Formula Weight 84.16
Percent Purity 99%
Chemical Name or Material trans-2-Hexene
Show More Show Less
Hazard Category H225-H304-H315-H319
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 223-752-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit